Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRB10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BN3
75934-452EA 518 USD InStock
75934-452
Anti-GRB10 Rabbit Polyclonal Antibody
Antibodies
GRB10 belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules and function in diverse cellular processes. GRB10 is a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. Overexpression of some isoforms of GRB10 inhibits tyrosine kinase activity and results in growth suppression. GRB10 can also interact with the L isoform of the proapoptotic protein Bim. This gene is imprinted in a highly isoform- and tissue-specific manner, with expression observed from the paternal allele in the brain, and from the maternal allele in the placental trophoblasts.

Anti-GRB10 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 67 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: GRB10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty