Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IdU Mouse Monoclonal Antibody [Clone: 32D8.D9]

Supplier: Rockland Immunochemicals
200-301-H51
75934-624EA 533 USD InStock
75934-624
Anti-IdU Mouse Monoclonal Antibody [Clone: 32D8.D9]
Antibodies
Iododeoxyuridine (5-Iodo-2'-deoxyuridine, IdU) is a synthetic thymidine nucleoside analog. IdU, like BrdU thymidine analog, is used to allow the detection of growing or proliferating cells in living tissues. During the S-phase of cell division, DNA replication occurs, and IdU can be incorporated into the newly synthesized DNA by substituting for naturally occurring thymidine. Antibodies specific for IdU are subsequently used to detect the incorporated IdU thymidine analog. This highlights cells that were actively replicating their DNA and is suggestive of actively growing cells. Antibody binding usually requires the DNA to be denatured, typically by exposing the cells to acid or heat.

Anti-IdU Antibody has been tested as suitable for western blot, ELISA, and immunofluorescence microscopy. This antibody may be suitable for additional immunoassays including flow cytometry and immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect to detect incoporated IdU thymidine analog from replicated cells.

Type: Primary
Antigen: IdU
Clonality: Monoclonal
Clone: 32D8.D9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty