Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IKBKG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BU1
75934-640EA 518 USD InStock
75934-640
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
Nuclear factor kappa B (NF-kB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kB is associated with IkB proteins in the cell cytoplasm, which inhibit NF-kB activity. The IkB kinase (IKKa and IKKb) phosphorylates IkB and mediates NF-kB activation. A novel molecule in the IKK complex was recently identified and termed IKKgamma/NEMO/FIP3/IKKAP1. IKKgamma interacts with IKKb and is required for the activation of IKK complex and NF-kB by LPS, PMA, TNF, and IL-1 stimulation. FIP3 was also shown to bind to RIP and NIK and to mediate TNF-induced NF-kB activation.

Anti-IKK gamma Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 48 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty