Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LMNB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P62
75934-812EA 487 USD InStock
75934-812
Anti-LMNB1 Rabbit Polyclonal Antibody
Antibodies
Lamin-B1 is a protein that in humans is encoded by the LMNB1 gene. The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. This gene encodes one of the two B type proteins, B1. This antibody is suitable for researchers interested in chromatin antibody research.

Anti-Lamin B1 is useful for Immunohistochemistry -P, -F, Immunocytochemistry, and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~66.4 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: LMNB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty