Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NANOG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H03
75934-944EA 518 USD InStock
75934-944
Anti-NANOG Rabbit Polyclonal Antibody
Antibodies
NANOG antibody detects human NANOG. Expression of NANOG is required for the maintenance of pluripotency in epiblast and embryonic stem (ES) cells as well as for the ability to maintain ES self-renewal independently of LIF/Stat3. The role of NANOG in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 (also known as Oct-4), another germline-specific transcription factor, and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing NANOG, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. Anti-NANOG antibodies are ideal for investigators involved in stem cell research.

Anti-NANOG Antibody is suitable for use in E,WB, and IHC. Expect a band approximately ~34.6 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty