Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOTUM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DD7
75934-956EA 518 USD InStock
75934-956
Anti-NOTUM Rabbit Polyclonal Antibody
Antibodies
In Drosophila, the notum gene is regulated by the Wingless pathway and encodes a secreted hydrolase that modifies heparan sulfate proteoglycans. The mammalian homolog has been shown to be able to cleave glypicans and can release GPI-anchored proteins from the mammalian cell surface. Like the Drosophilia NOTUM, the mammalian protein can act as a negative regulator of the Wnt signaling pathway. NOTUM is expressed at a low level in most mammalian tissues, although it is overexpressed in a subset of human hepatocellular carcinomas. Its transcription is regulated by beta-catenin/TCF and is a target of the Wnt signaling pathway, forming a negative feedback loop that regulates the expression and activity of the Wnt pathway.

Anti-NOTUM Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NOTUM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty