Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYBB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DD9
75934-962EA 518 USD InStock
75934-962
Anti-CYBB Rabbit Polyclonal Antibody
Antibodies
The NOX family of NAPDH oxidases is comprised of seven transmembrane proteins that oxidize intracellular NAPDH/NADH, causing electron transport across the membrane and the reduction of molecular oxygen to superoxide (1). NOX2, also known as cytochrome b beta (CYBB) is one of two proteins that make up Cytochrome b-245, thought to be a primary component of the microbicidal oxidase system of phagocytes. NOX2 deficiency is one of five described biochemical defects associated with chronic granulomatous disease (CGD) (2). Activation of the NOX2 enzyme complex in microglia is thought to be neurotoxic and may play a role in Alzheimer’s and Parkinson’s disease (3).

Anti-NOX2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 65 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: CYBB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty