Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OLIG2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X18
75934-970EA 518 USD InStock
75934-970
Anti-OLIG2 Rabbit Polyclonal Antibody
Antibodies
Olig2 is a well conserved bHLH transcription factor that shows both anti-neural functions and pro-neural functions at different stages in the formation of the oligodendrocyte lineage. Olig2 is expressed in 100% of the human diffuse gliomas irrespective of grade and required for intracranial tumor formation in a genetically relevant model of malignant glioma. A developmentally regulated triple serine motif at positions 10, 13 and 14 in the amino terminus is well conserved across species ranging from humans to zebrafish and is essential for Olig2 proliferative function in both normal and malignant neural progenitors. All three serine residues must be mutated to achieve a strong loss-of-function or gain-of-function phenotype, suggesting that the phosphorylation state of Olig2 represents a significant conformational change in the amino terminus. Olig2 pS 10/13/14 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser10,13,14 Olig2 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~32 kDa Olig2 phosphorylated at Ser10, Ser13, and Ser14. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: OLIG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty