Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PION Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H11
75934-984EA 518 USD InStock
75934-984
Anti-PION Rabbit Polyclonal Antibody
Antibodies
Pion antibody detects human PION. Accumulation of the amyloid-beta peptide (Abeta) in the cerebral cortex is a critical event in the pathogenesis of Alzheimer’s disease. The beta−amyloid protein precursor (APP) is cleaved by one of two beta−secretases (BACE and BACE2), producing a soluble derivative of the protein and a membrane anchored 99 -amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for gamma−secretase to generate the 4 kDa amyloid-beta peptide (Abeta), which is deposited in the Alzheimer’s disease patients’ brains. PION, or GSAP, selectively increases amyloid-beta production through a mechanism involving its interaction with both gamma-secretase and the APP C-terminal fragment, suggesting that PION may be a potential therapeutic target for the treatment of Alzheimer’s disease. Anti-PION antibodies are ideal for investigators involved in neuroscience research.

Anti-PION Antibody is suitable for use in E,WB, and IHC. Expect a band approximately ~97.8 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: PION
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty