Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLAUR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P91
75935-004EA 487 USD InStock
75935-004
Anti-PLAUR Rabbit Polyclonal Antibody
Antibodies
PLAUR(PLASMINOGEN ACTIVATOR RECEPTOR, UROKINASE-TYPE), also known as UPAR or CD87, is multidomain glycoprotein tethered to the cell membrane with a glycosylphosphotidylinositol (GPI) anchor. PLAUR consists of three different domains of the Ly-6/uPAR/alpha-neurotoxin family. PLAUR is originally identified as a saturable binding site for urokinase on the cell surface. And the gene plays an important role in many normal as well as pathologic processes. The PLAUR gene is localized to 19q13.31. PLAUR is a part of the plasminogen activation system, which in the healthy body is involved in tissue reorganization events such as mammary gland involution and wound healing. PLAUR binds urokinase and thus restricts plasminogen activation to the immediate vicinity of the cell membrane. Thus it seems to be an important player in the regulation of this process. In human coronary artery vascular smooth muscle cells, UPA stimulates cell migration via a UPAR signaling complex containing TYK2 and phosphatidylinositol 3-kinase. This antibody is suitable for researchers interested in neuroscience research.

Anti-PLAUR is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~37 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: PLAUR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty