Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RASSF10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DW7
75935-146EA 518 USD InStock
75935-146
Anti-RASSF10 Rabbit Polyclonal Antibody
Antibodies
RASSF10 is a member of the N-terminal Ras association domain-containing family of proteins, a family of tumor suppressor genes that are frequently epigenetically inactivated in cancer. These proteins consist of RASSF7-10 and are structurally distinct from RASSF1-6, containing the N-terminal Ras association domain but lacking the Sav/RASSF/Hippo protein interaction domains. While little is known of the function of RASSF10, it is inactivated in both childhood leukemia and gliomas. In secondary glioblastomas, RASSF10 methylation can be used as an independent prognostic associated with worst progression-free survival and overall survival.

Anti-RASSF10 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 57 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RASSF10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty