Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOXA Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-093S
75935-234EA 148 USD InStock
75935-234
Anti-SOXA Goat Polyclonal Antibody (BAC)
Antibodies
Sarcosine oxidase is an inducible enzyme that catalyzes the oxidative demethylation of sarcosine to glycine, hydrogen peroxide, and 5,10-methylene-tetrahydrofolate in the presence of tetrahydrofolate and oxygen. Sarcosine oxidase is also able to oxidize cyclic amino acids such as L-proline and L-pipecolic acid, albeit at reduced rates.

Anti-sarcosine oxidase antibody has been assayed against 1.0 ug of Sarcosine Oxidase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.

Type: Primary
Antigen: SOXA
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty