Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SESN2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EG9
75935-240EA 518 USD InStock
75935-240
Anti-SESN2 Rabbit Polyclonal Antibody
Antibodies
SESTRIN2, also known as Hi95, is a member of the sestrin family of PA26-related proteins and is induced following DNA damage or oxidative stress (1). SESTRIN2 and the related protein SESTRIN1 interact with the NRF2 suppressor KEAP1, the autophagy substrate p62 and the ubiquitin ligase RBX1 to protect cells against oxidative stress by activating NRF2 and promoting the p62-dependent autophagic degradation of KEAP1 (2). SESTRIN2 also negatively regulates TOR signaling by activating AMP-activated protein kinase (AMPK) and TSC2 phosphorylation (3), thereby protecting cells against energetic stress-induced death (4).

Anti-SESTRIN2 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 54 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SESN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty