Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SETD7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V83
75935-242EA 487 USD InStock
75935-242
Anti-SETD7 Rabbit Polyclonal Antibody
Antibodies
Set9 is a histone methyltransferase which specifically methylates lysine 4 of histone H3 being a specific tag for epigenetic transcriptional activation. Further, methylation of histone H3K4 by Set9 precludes deactylation of H3 by the NuRD complex and impairs Suv39h1-mediated methylation of H3K9. Set9 also methylates the tumor suppressor p53 at lysine 372, thereby enchancing its stability. IT contains a SET domain but lacks the pre- and post-SET domains. Anti-Set9 Antibody is ideal for research in Epigenetics, Gene Expression and Chromatin Remodeling.

Anti-Set9 Antibody is suitable for Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 50 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: SETD7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty