Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SETD1A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V94
75935-244EA 487 USD InStock
75935-244
Anti-SETD1A Rabbit Polyclonal Antibody
Antibodies
SETD1A is a component of the SET1 histone methyltransferase (HMT) complex. This complex specifically methylates lysine 4 of histone H3, but only if the neighboring lysine 9 residue is not yet methylated. Methylation of H3K4 represents a specific tag for epigenetic transcriptional activation. SETD1A shows a non-overlapping localization with SETD1B, suggesting that both proteins play a specific role in the epigenetic control of chromatin structure and gene expression. Anti-Setd1a Antibody is ideal for research in Epigenetics and Gene Expression.

Anti-Setd1a Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 186 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: SETD1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty