Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC10A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P84
75935-380EA 487 USD InStock
75935-380
Anti-SLC10A1 Rabbit Polyclonal Antibody
Antibodies
SLC10A1(SOLUTE CARRIER FAMILY 10 (SODIUM/BILE ACID COTRANSPORTER FAMILY), MEMBER 1),also called NTCP or NTCP1,is found in the basolateral membranes of hepatocytes, which is also involved in the reabsorption of bile acids. The SLC10A1 gene is mapped on 14q24.2.The SLC10A1 gene contains 5 exons and spans about 23 kb. Like other sodium/bile acid transporters, SLC10A1 contains an exoplasmic N terminus, an odd number of transmembrane regions, and a cytoplasmic C terminus. Two amphipathic sequences are critical for intramolecular interactions and for proper trafficking of SLC10A1 to the plasma membrane.Injection of in vitro transcribed NTCP cRNA into Xenopus oocytes resulted in the expression of Na(+)-dependent taurocholate uptake. NTCP-mediated taurocholate uptake in oocytes was inhibited by all major bile acid derivatives and by bromosulfophthalein. This antibody is suitable for researchers interested in cancer research.

Anti-SLC10A1 is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~38.1kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: SLC10A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty