Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC1A7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EN8
75935-382EA 518 USD InStock
75935-382
Anti-SLC1A7 Rabbit Polyclonal Antibody
Antibodies
The solute carrier family 1 member 7 (SLC1A7) protein was initially identified as a glutamate transporter coupled to a chloride conductance that is expressed primarily in the retina (1). More recent evidence indicates that is it widely expressed in peripheral tissues, also (2). Experiments have shown that SLC1A7 functions as a low-affinity/low-capacity glutamate transport, with an anion channel optimized for anion conductance in the negative voltage range (3). In the retina, SLC1A7 co-localizes with the serum and glucocorticoid dependent kinase SGK1, and its cell surface expression and activity is regulated SGK1 and its isoform SGK3 (4).

Anti-SLC1A7 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 61 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SLC1A7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty