Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STIM1 Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-601-J49
75935-400EA 518 USD InStock
75935-400
Anti-STIM1 Sheep Polyclonal Antibody
Antibodies
Stim I antibody helps regulate calcium influx after the depletion of intracellular calcium stores. It functions as a calcium sensor in the endoplasmic reticulum via its EF-hand domain. After depletion of Ca+2, it is translocated from the endoplasmic reticulum to the plasma membrane so the calicum release-activated channel subunit is activated. Anti-Stim I antibody is ideal for investigators interested in Metabolism and Signal Transduction research.

Anti-Stim I antibody is useful for ELISA, immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~77kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: STIM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty