Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TESPA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FC7
75935-438EA 518 USD InStock
75935-438
Anti-TESPA1 Rabbit Polyclonal Antibody
Antibodies
TESPA1 is a critical component of the TCR signalosome and is essential for T cell selection and maturation through the regulation of TCR signaling during T cell development (1). TESPA1 encodes a protein of 458 amino acids and is highly conserved among vertebrate species (2). TESPA1 expression was tightly regulated during T cell development, with highest expression at the DP stage (3). TESPA1 deficiency resulted in failure of positive selection and substantially fewer of CD4+ and CD8+ SP cells in the thymus (1,4).

Anti-TESPA1 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 59 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TESPA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty