Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TET1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FC8
75935-440EA 518 USD InStock
75935-440
Anti-TET1 Rabbit Polyclonal Antibody
Antibodies
TET1, a member of the ten-eleven-translocation (TET) family of genes, was initially discovered as a fusion partner of MLL in acute myeloid leukemias containing the t(10;11)(q22;q23) (1). It is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine (2). TET1 expression is regulated during inflammation in both THP-1 and primary dendritic cells, and is a negative transcriptional regulator of IL-1beta following and inflammatory stimulus (3). Recent evidence has shown that TET1 is part of a signaling pathway that regulates breast cancer growth and metastasis (4).

Anti-TET1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 235 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TET1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty