Would you like to log in again?
SPE CARTRIDGE FLORSL SILIAPREP 50MG/ 1ML
Supplier: SiliCycle
SiliaPrep* SPE cartridge Florisil 50 mg^ SiliaPrep* SPE cartridge, Florisil, Particle size: 75 - 150 um, Pore Size: 100 A, Bed Weight: 50 mg, Column volume: 1 mL, A polar sorbent (MgO3Si) presenting a basic character used to extract non-polar to moderately polar compounds from non-polar solvents
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE FLORSL SILIAPREP 50MG/ 1ML 75957-726 EAUT001401B Box of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty