Would you like to log in again?
SPE CARTRIDGE C18 WPD SILIAPREP 200 MG
Supplier: SiliCycle
SiliaPrep* SPE cartridge C18 WPD 200 mg^ SiliaPrep* SPE cartridge, C18 WPD (Widepore), Particle size: 37 - 55 um, Pore Size: 125 A, Bed Weight: 200 mg, Column volume: 3 mL, Carbon Load: 13 %, With end capping, pH Stability: 3 - 8, Moderatly hydrophobic, non-polar and highloading capacity sorbent
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE C18 WPD SILIAPREP 200 MG 75957-934 ER33229G03G Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty