Would you like to log in again?
SPE CARTRIDGE WCX SILIAPREPX 200MG BX-50
Supplier: SiliCycle
SiliaPrepX* SPE cartridge WCX 200mg^ SiliaPrepX* SPE cartridge, WCX (DVB copolymer functionalized with carboxylic acid), Particle size: 85 um, Pore Size: 60 A, Bed Weight: 200mg, Column volume: 3mL, Carbon load: 85%, Ph Stability: 1 - 14, Applications: Strong basic compounds, Streptomycin from food
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE WCX SILIAPREPX 200MG BX-50 75958-068 EP001503G Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty