Would you like to log in again?
SPE CARTRIDGE PRS SCX-2 NEC SILIAPREP 1G
Supplier: SiliCycle
SiliaPrep* SPE cartridge Propylsulfonic Acid (SCX-2) nec 1 g^ SiliaPrep* SPE cartridge Propylsulfonic Acid (SCX-2) nec, Particle size: 40-63 um / 230-400 mesh, Pore Size: 60 A, Bed Weight: 1 g, Column volume: 6 mL, is a strong cation exchanger and maintains a negative charge throughtout the pH scale
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE PRS SCX-2 NEC SILIAPREP 1G 75958-526 ER51430B06S Box of 50 (1g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty