Would you like to log in again?
SPE CARTRIDGE AM WAX SILIAPREP 50MG/ 1ML
Supplier: SiliCycle
SiliaPrep* SPE cartridge Amine (WAX) 50 mg^ SiliaPrep* SPE cartridge, Amine (WAX), Particle size: 40-63 um / 230-400 mesh, Pore Size: 60 A, Bed Weight: 50 mg, Column volume: 1 mL, is a weak anion exchanger sorbent used in replacement of SAX for strong anions to avoid irreversible retention
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE AM WAX SILIAPREP 50MG/ 1ML 75958-532 ER52030B01B Box of 100 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty