Would you like to log in again?
SPE CARTRIDGE IMIDAZOLE SILIAPREP 50MG
Supplier: SiliCycle
SiliaPrep* SPE cartridge Imidazole 50 mg^ SiliaPrep* SPE cartridge Imidazole, Particle size: 40-63 um, Pore Size: 60 A, Bed Weight: 50 mg, Column volume: 1 mL, Metal Scavenger, Lower the residual metal concentration of various metal complexes (Pd, Pt, Rh, Ru, Ni, Sn, etc) to single digit ppm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE IMIDAZOLE SILIAPREP 50MG 75958-920 ER79230B01B Box of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty