Would you like to log in again?
SPE CARTRIDGE CYSTEINE SILIAPREP 200MG
Supplier: SiliCycle
SiliaPrep* SPE cartridge Cysteine 200 mg^ SiliaPrep* SPE cartridge Cysteine, Particle size: 40-63 um / 230-400 mesh, Pore Size: 60 A, Bed Weight: 200 mg, Column volume: 3 mL, Lower the residual metal concentration of various metal complexes (Pd, Pt, Rh, Ru, Ni, Sn, etc) to single digit ppm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPE CARTRIDGE CYSTEINE SILIAPREP 200MG 75958-940 ER80530B03G Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty