Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MFGE8 Mouse Monoclonal Antibody [clone: EDM45]

Supplier: Biotium

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


BNUB0942-100 BNUB0942-500
75961-250EA 499.07 USD InStock
75961-250 75961-248
Anti-MFGE8 Mouse Monoclonal Antibody [clone: EDM45]
Antibodies
This antibody recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: Clone EDM45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty