Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NAPSA Mouse Monoclonal Antibody [clone: NAPSA/1239]

Supplier: Biotium

BNUB1239-500 BNUB1239-100
75965-504EA 499.07 USD InStock
75965-504 75965-502
Anti-NAPSA Mouse Monoclonal Antibody [clone: NAPSA/1239]
Antibodies
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.

Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: Clone NAPSA/1239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty