Would you like to log in again?
BioClean™ Emerald Sterile Nitrile Cleanroom Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioClean™
BENS10.0 BENS9.0 BENS9.0 BENS8.5 BENS8.0 BENS6.0 BENS7.5 BENS7.0 BENS7.0 BENS6.5 BENS7.5 BENS6.0 BENS6.5 BENS8.0 BENS10.0 BENS8.5
75972-648CS 770.74 USD InStock
CA75972-648 CA75972-588 75972-648 75972-590 CA75972-590 75972-592 CA75972-586 75972-586 CA75972-584 75972-588 75972-594 CA75972-582 CA75972-594 75972-582 CA75972-592 75972-584
BioClean™ Emerald Sterile Nitrile Cleanroom Gloves, Ansell
Gloves
Gloves are resistant to a range of chemicals, can be easily double-donned, and are accelerator-free to reduce the risk of latex allergies. The hand specific shape ensures wearer comfort and reduces hand fatigue, while enabling good dexterity.
  • Powder, sulfur and accelerator free
  • Textured finish with a smooth feel
  • Anti-static, no natural latex proteins
  • Internal glove surface is powder-free and chlorinated
Sterilized by gamma radiation, easy double-donning.
Certifications: ISO 9001: 2008 facility certifications. Conforms to Category 3. Processed in an ISO Class 4 environment. Complies with EN 420:2003, EN 374-1, 2 & 3:2003, and EN 455-2:2009 glove standards. Permeation standard ASTM D6978-05 for cytotoxic drugs. Sterilized in accordance with ISO 11137-1:2006. Suitable for contact with all foodstuffs. CE 0120 marked. EU GMP Grade A compatible.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty