Would you like to log in again?
BioClean-D™ Sterile Coveralls with Hood and Integrated Boots, Ansell
Supplier: Ansell
BioClean-D™
S-BDFC-L S-BDFC-XL S-BDFC-XXXXL S-BDFC-M S-BDFC-XXL S-BDFC-XXL S-BDFC-S S-BDFC-XXXL S-BDFC-XXXL S-BDFC-L S-BDFC-M S-BDFC-XL S-BDFC-XXXXL S-BDFC-S
75982-212CS 587.26 USD InStock
CA75982-210 75982-212 75982-210 75982-204 CA75982-214 CA75982-202 75982-202 CA75982-212 75982-214 75982-208 CA75982-206 75982-206 CA75982-204 CA75982-208
BioClean-D™ Sterile Coveralls with Hood and Integrated Boots, Ansell
Disposable Garments
The BioClean-D™ coverall with hood and integrated boots is a disposable garment offering comfort and head-to-toe protection.
  • Anti-static, lightweight, low-linting CleanTough™ material construction
  • Thumb loops to ensure a secure hold
  • Front zip with sealable cover and protective flap
  • Elasticated back, cuffs, and ankles
  • Slip resistant soles
  • Excellent ESD properties
  • Latex-free
The integrated boots feature slip-resistant polyurethane soles to ensure every step can be taken with confidence. ISO Class 4 cleanroom compatible. These sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739, EN1149-5, CATEGORY III, Sterilized in accordance with ISO 11137-1:2006.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty