Would you like to log in again?
BioClean-D™ Sterile Extra-Length Overboots, Ansell
Supplier: Ansell
BioClean-D™
BDOB BDOB S-BDOB S-BDOB
75982-216CS 747 USD InStock
75982-216 CA75972-644 75972-644 CA75982-216
BioClean-D™ Sterile Extra-Length Overboots, Ansell
Disposable Garments
These disposable boot covers are made of BioClean-D™, a lightweight, low linting and hard-wearing fabric (PP laminated with a PE film) which provides maximum comfort and protects against fine sprays and particles.
  • Longer length: 500 mm
  • Low-linting
  • Tie-fastenings at top and ankle
  • Slip-resistant polyurethane sole
  • Latex-free
  • Available as sterile or nonsterile version
  • Elasticated top with secure ties
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Overboots feature a slip resistant sole and easy tie-fastenings at the top and ankle. Suitable for use in Class 10, ISO 4, M2.5 environments. Sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Packaging: Sterile version: One pair per sealed plastic bag, 15 sealed bags per sealed outer bag, five sealed bags of 15 pairs per sealed carton liner, one carton liner per carton (75 pairs).
Nonsterile version: 5 bags of 30 pieces per sealed carton liner; one carton liner per carton (150 pieces).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty