Would you like to log in again?
BioClean-D™ Sterile Coveralls with Hood, Ansell
Supplier: Ansell
BioClean-D™
S-BDCHT-L S-BDCHT-XXL S-BDCHT-M S-BDCHT-XXXL S-BDCHT-XS S-BDCHT-L S-BDCHT-XXL S-BDCHT-S S-BDCHT-XXXXL S-BDCHT-XL S-BDCHT-XXXL S-BDCHT-XL S-BDCHT-XXXXL S-BDCHT-S S-BDCHT-M
75982-276CS 512.16 USD InStock
CA75982-276 CA75982-274 CA75982-284 75982-276 CA75982-278 75982-280 75982-270 CA75982-272 75982-284 CA75982-282 75982-274 75982-282 CA75982-270 CA75982-280 75982-272
BioClean-D™ Sterile Coveralls with Hood, Ansell
Disposable Garments
The BioClean-D™ coverall with hood is a disposable garment.
  • Lightweight CleanTough™ material
  • Thumb loops to ensure a secure hold
  • Front zip with sealable cover and protective flap
  • Elasticated hood, back, cuffs, and ankles
  • Excellent ESD properties
  • Latex-free
The anti-static, low-linting CleanTough™ material provides comfort and protection from a range of chemicals. Lightweight yet durable garments feature bound seams with single needle stitching. ISO Class 4 cleanroom compatible. These sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739, EN ISO 13688:2013, EN 1 ASTM F739, EN ISO 13688:2013, EN 14325.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty