Would you like to log in again?
Multi-Purpose Containers with Snap Cap, Globe Scientific
Supplier: Globe Scientific
271009W 271064W 271172W 271032W 271086W 271008W 271016W
75982-652CS 163.68 USD InStock
75982-652 75982-650 75982-656 75982-654 75982-648 75982-646 75982-658
Multi-Purpose Containers with Snap Cap, Globe Scientific
Sample Containers General Purpose Containers
These disposable containers are produced from high density polyethylene and are ideal for the collection and storage of histology and pathology specimens, fluids, powders, and solids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty