Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pipette Tips, Watson Bio Lab
Supplier: Watson Bio Lab
120-805CS 120-805C 122-204CS 1272-801CS 120-801C 120-207C 123R-757CS 123R-254CS 124-1200S 126-1000S 124-5000S 120-401CS 110-805C 124P-20S 120-401C 1271-204CS 1251P-204CS 126-1200S 122-805CS 120P-204CS 126-20S 122-207CS 1252-801CS 124P-100S 110-705C 123R-755CS 126-5000S 110-207C 120-204C 110-801C 122-801CS 1272-703CS 126-100S 1252-703CS 120P-207CS 113-743C 110-204C 1201-705CS 1272-207CS 1201-705C 124P-1000S 120-801CS 1252P-207CS
75982-784BX 229.39 USD InStock
75982-784 75982-782 75982-760 75982-820 75982-766 75982-742 76191-290 75982-758 76191-292 75982-836 75982-814 75982-734 75982-778 75982-834 75982-812 75982-818 75982-738 75982-838 75982-816 75982-750 75982-772 76191-300 75982-770 75982-776 75982-832 75982-810 75982-774 75982-730 75982-752 75982-830 75982-808 75982-802 75982-824 75982-748 75982-822 75982-768 75982-746 75982-828 75982-806 76191-286 75982-804 75982-826 75982-728
Pipette Tips, Watson Bio Lab
Pipette Tips
Watson offers pipette tips in a wide variety of volumes, lengths, and widths.
  • Thin-walled tips available
  • Soft-fitting tips available
  • Tips come in short, standard, long, and extra-long sizes
  • Large bore tips available
  • Graduated tips provide simple measurement
Accessories information: Empty racks are available separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty