Would you like to log in again?
Screwcap Tubes without Caps, Watson Bio Lab
Supplier: Watson Bio Lab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
139-120-111C 139-120-112C 139-115-111BL 139-120-111BL 139-115-112C 139-105-111BL
75982-908PK 79.39 USD InStock
75982-908 75982-912 75982-910 75982-916 75982-906 75982-914
Screwcap Tubes without Caps, Watson Bio Lab
Tubes Test Tubes
These tubes use medical-grade polypropylene to provide resistance against a centrifugal force as high as 30000×g.
  • Flat top with marking space
Tubes feature a marker, or notch, on the flange of the tube, which helps position the tube during centrifugation. The silicone rubber O-ring is resistant to pressure and is suitable for storage in liquid nitrogen (vapor phase) or a deep freezer.
Accessories information: Screwcaps available separately.
Caution: Store liquid nitrogen in the vapor phase for safety reasons. In the liquid phase, liquid nitrogen may flow into the tube, resulting in tube rupture and the nitrogen spatter during defrosting. Wear a face shield, protective clothes, and gloves when handling liquid nitrogen for defrosting.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty