Would you like to log in again?
RACK STRG FRZR PC RED 131X131X52MM 81TUB
Supplier: Watson Bio Lab
Freezer Storage Rack 131x131x52mm^ Freezer Storage Rack, Material: Polycarbonate, Color: Red, Heatproof and crashproof, It is possible to use it for the liquid nitrogen (vapor phase) and the deep freezer preservation, For 81 tubes, Dimension: 131x131x52mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK STRG FRZR PC RED 131X131X52MM 81TUB 75982-940 1577-850R Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty