Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
PCR Tubes, Watson Bio Lab
Supplier: Watson Bio Lab
137-333C 137-431C 137-232C 137-212C 137-231C 137-432C 137-211C
75982-960CS 4882.75 USD InStock
75982-960 75982-952 75982-964 75982-962 75982-956 75982-954 75982-958
PCR Tubes, Watson Bio Lab
Tubes PCR Tubes
The side walls are uniformly thin, which enables PCR with superior heat exchange.
  • Thin wall, high transparency
  • Nuclease-free certified
Flat and dome caps are available.
All tubes feature autoclavable polypropylene construction.
The flat top, smooth and superior in optical transparency, is the most suitable for Real Time PCR. They also offer writing space.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty