Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Fibrinogen Goat Polyclonal Antibody (Horseradish Peroxidase )

Supplier: MP Biomedicals
0855239
75983-042EA 629.3 USD InStock
75983-042
Anti-Fibrinogen Goat Polyclonal Antibody (Horseradish Peroxidase )
Antibodies
Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to human fibrinogen and buffer salts.

The protein encoded by FGA gene is the alpha component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia, afibrinogenemia and renal amyloidosis. Alternative splicing results in two isoforms which vary in the carboxy-terminus.

Dot Blot, Immunomicroscopy

Type: Primary
Antigen: Fibrinogen
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty