Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C3 Goat Polyclonal Antibody (Fluorescein)

Supplier: MP Biomedicals
0855510
75983-056EA 1058.34 USD InStock
75983-056
Anti-C3 Goat Polyclonal Antibody (Fluorescein)
Antibodies
The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. The F(ab')2 fragments are tested for purity and specificity at a minimum of 20 mg/mL using immunoelectrophoresis; No trace of albumin, whole molecule IgG or Fc is detected. Antibody activity to other serum proteins is not present.

Immunoblotting

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty