Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bovine Serum Goat Polyclonal Antibody

Supplier: MP Biomedicals
0855998
75983-072EA 345.19 USD InStock
75983-072
Anti-Bovine Serum Goat Polyclonal Antibody
Antibodies
Product is the lyophilized powder of goat antiserum to human serum and buffer salts.
The antiserum is prepared by immunizing goats with human serum; antiserum is delipidated and made specific by
immunoadsorption with soluble antigens or solid-phase immunoadsorption where appropriate. The resulting antiserum is
dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer,
filtered through a 0.22 µm filter, vialed and lyophilized

Type: Primary 
Antigen: Bovine Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Cat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty