Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Albumin Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
0856963
75983-074EA 537.35 USD InStock
75983-074
Anti-Albumin Rabbit Polyclonal Antibody
Antibodies
The total protein is measured by absorbance (A280nm). Antibody titer is standardized by equivalence-point precipitation. Each affinity purified antibody is tested for purity at 10 mg/mL using immunoelectrophoresis. The product is rabbit IgG; no trace of albumin is detected.

Immunoassays

Type: Primary
Antigen: Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty