Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaBond® C4, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R32030B-5G R32130B-100G R32130B-1KG R32030B-1KG R32030B-1KG R32030B-250G R32130B-500G R32130B-1KG R32130B-50G R32030B-100G R32030B-10G R32130B-5G R32130B-250G R32030B-500G R32130B-100G R32130B-10G R32130B-50G R32030B-50G R32030B-500G R32030B-50G R32030B-250G R32030B-25G R32130B-250G R32030B-5G R32130B-25G R32130B-10G R32030B-100G R32030B-25G R32130B-25G R32130B-500G R32030B-10G R32130B-5G
75991-230EA 179.48 USD InStock
CA75991-218 75991-230 75991-232 CA75991-240 75991-234 75991-212 CA75991-242 CA75991-220 75991-236 75991-214 CA75991-222 75991-216 75991-238 CA75991-224 75991-218 CA75991-226 CA75991-228 75991-240 CA75991-230 75991-220 75991-242 CA75991-232 75991-222 CA75991-212 CA75991-234 75991-224 CA75991-214 CA75991-236 75991-226 CA75991-216 CA75991-238 75991-228
SiliaBond® C4, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty