Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaBond® C2, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R32630B-5G R32530B-500G R32630B-1KG R32530B-5G R32630B-500G R32530B-25G R32530B-1KG R32630B-5G R32630B-50G R32530B-50G R32530B-100G R32530B-500G R32530B-10G R32530B-100G R32630B-25G R32630B-10G R32530B-5G R32530B-1KG R32530B-50G R32630B-1KG R32630B-250G R32630B-100G R32530B-250G R32630B-25G R32630B-500G R32530B-25G R32530B-10G R32530B-250G R32630B-10G R32630B-100G R32630B-250G R32630B-50G
75991-270EA 3914.91 USD InStock
CA75990-610 75991-270 75991-250 75991-272 75991-252 CA75991-260 75991-254 CA75991-262 75991-256 CA75991-264 75991-258 CA75991-244 CA75991-266 CA75991-268 CA75991-246 CA75991-248 75990-610 CA75991-270 75991-260 CA75991-272 CA75991-250 75991-262 CA75991-252 75991-264 CA75991-254 75991-244 75991-266 CA75991-256 75991-246 75991-268 CA75991-258 75991-248
SiliaBond® C2, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty