Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaBond® C18, 17% Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R33230B-1KG R33230B-10G R33330B-100G R33330B-5G R33230B-100G R33330B-5G R33330B-25G R33230B-50G R33230B-5G R33230B-250G R33330B-10G R33230B-10G R33230B-500G R33330B-100G R33330B-10G R33230B-100G R33230B-1KG R33330B-500G R33230B-5G R33330B-25G R33230B-250G R33330B-50G R33230B-25G R33330B-1KG R33330B-1KG R33330B-250G R33230B-500G R33230B-50G R33330B-250G R33330B-50G R33230B-25G R33330B-500G
75991-308EA 188.89 USD InStock
75991-308 CA75991-316 CA75991-318 75991-330 CA75991-320 75991-310 CA75991-300 CA75991-322 75991-312 CA75991-302 CA75991-324 75991-314 CA75991-304 CA75991-326 75991-316 CA75991-306 CA75991-328 75991-318 CA75991-308 75991-320 75991-300 75991-322 CA75991-330 75991-302 75991-324 CA75991-310 75991-304 75991-326 CA75991-312 75991-306 75991-328 CA75991-314
SiliaBond® C18, 17% Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty