Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaBond® Glycidoxy, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R36030B-25G R36030B-50G R36030B-5G R36030B-25G R36030B-250G R36030B-1KG R36030B-100G R36030B-500G R36030B-1KG R36030B-10G R36030B-500G R36030B-10G R36030B-5G R36030B-100G R36030B-50G R36030B-250G
75991-418EA 1674.51 USD InStock
CA75991-416 75991-418 CA75991-418 75991-408 CA75991-408 75991-410 75991-420 75991-412 CA75991-410 75991-422 CA75991-420 75991-414 CA75991-412 CA75991-422 75991-416 CA75991-414
SiliaBond® Glycidoxy, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty