Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaBond® Propylsulfonic Acid, SCX-2, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R51430B-250G R51430B-100G R51430B-25G R51430B-500G R51430B-10G R51430B-1KG R51230B-5G R51430B-1KG R51230B-5G R51430B-5G R51230B-500G R51230B-100G R51430B-25G R51230B-50G R51430B-10G R51230B-100G R51430B-500G R51430B-50G R51430B-100G R51230B-1KG R51430B-5G R51230B-50G R51230B-25G R51230B-250G R51230B-250G R51230B-10G R51230B-500G R51230B-25G R51430B-250G R51430B-50G R51230B-10G R51230B-1KG
75991-528EA 107.1 USD InStock
75991-528 CA75991-536 CA75991-538 75991-550 CA75991-540 75991-552 75991-530 CA75991-542 75991-554 75991-532 CA75991-544 75991-556 75991-534 CA75991-546 75991-536 CA75991-526 CA75991-548 75991-538 CA75991-528 75991-540 75991-542 CA75991-550 75991-544 CA75991-552 CA75991-530 75991-546 CA75991-532 CA75991-554 75991-548 75991-526 CA75991-556 CA75991-534
SiliaBond® Propylsulfonic Acid, SCX-2, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty