Would you like to log in again?
SiliaBond® Carbonate, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R66030B-100G R66030B-5G R66030B-100G R66030B-50G R66030B-25G R66030B-10G R66030B-250G R66030B-250G R66030B-500G R66030B-1KG R66030B-25G R66030B-1KG R66030B-5G R66030B-10G R66030B-50G R66030B-500G
75992-318EA 754.49 USD InStock
CA75991-768 CA75992-318 75992-318 CA75992-324 75992-324 CA75992-322 CA75992-320 75992-322 75992-320 75991-770 75991-772 CA75991-770 75991-774 CA75991-772 CA75991-774 75991-768
SiliaBond® Carbonate, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty