Would you like to log in again?
MULTIVIEW® PLUS (mouse-HRP/rabbit-AP) IHC Kit (Brown/Green), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT181-0150
75993-474EA 819 USD InStock
75993-474
MULTIVIEW® PLUS (mouse-HRP/rabbit-AP) IHC Kit (Brown/Green), Enzo Life Sciences
Assays Immunological Assays
Superior multiplex IHC kit for  efficient detection of multiple antigens in a single tissue sample.
  • Biotin-free nanopolymer-based system for superior multiplex antigen detection
  • Complete kit includes HIGHDEF® Chromogens for sensitive, crisp 2-color development
  • Preserves precious tissue while reducing reagent and labor costs
  • Expandable with additional chromogens (sold separately)
The MULTIVIEW® PLUS (mouse-HRP/rabbit-AP) IHC Kit is a non-biotin one-step detection system suitable for detecting and localizing antigens in formalin-fixed paraffin-embedded tissues.
Contains Antigen Retrieval Reagent, pH 9 (10X), ISH/IHC Peroxidase Block, Antibody Blocker/Diluent, MULTIVIEW® PLUS HRP (Anti-Mouse) Reagent, MULTIVIEW® PLUS AP (Anti-Rabbit) Reagent, HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate, HIGHDEF® Green AP Chromogen, HIGHDEF® Green AP Substrate, Hematoxylin, Empty Mixing Bottle.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty