Would you like to log in again?
Nitrate Substrate Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z124 R97 Z125
75993-554PK 99.49 USD InStock
75993-554 75993-560 75993-558
Nitrate Substrate Broth, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
For the determination of nitrate reactions of mycobacteria. Use with AFB Nitrate Reagent A and B
Reagent A contains sulfanilimide, 0.2%.
Reagent A contains sulfanilimide, 0.2%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty